Order Entry
Ireland
ContactUsLinkComponent
(N-Succinimidyloxycarbonylmethyl)tris(2,4,6-trimethoxyphenyl)phosphonium bromide acylation reagent, for protein sequencing by MALDI-MS, Supelco®
(N-Succinimidyloxycarbonylmethyl)tris(2,4,6-trimethoxyphenyl)phosphonium bromide acylation reagent, for protein sequencing by MALDI-MS, Supelco®
Catalog # 29208-50MG
Supplier:  Merck
CAS Number:  
(N-Succinimidyloxycarbonylmethyl)tris(2,4,6-trimethoxyphenyl)phosphonium bromide acylation reagent, for protein sequencing by MALDI-MS, Supelco®
Catalog # 29208-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :768.54 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:226409-58-1
Formula:C₃₃H₃₉BrNO₁₃P
MDL Number:MFCD03095505