Order Entry
Ireland
ContactUsLinkComponent
N-tert-Butyl-ɑ-phenylnitrone ≥99.5% (by HPLC) for ESR-spectroscopy, Supelco®
N-tert-Butyl-ɑ-phenylnitrone ≥99.5% (by HPLC) for ESR-spectroscopy, Supelco®
Catalog # 80126-1G
Supplier:  Merck
CAS Number:  
undefined
N-tert-Butyl-ɑ-phenylnitrone ≥99.5% (by HPLC) for ESR-spectroscopy, Supelco®
Catalog # 80126-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :177.25 g/mol
Storage Temperature:Freezer
CAS Number:3376-24-7
Formula:C₁₁H₁₅NO
MDL Number:MFCD00008799