Methanesulphonic acid 0.1 M (0.1N) in water eluent concentrate for IC, Supelco®
Catalog # 55517-1L
Supplier: Merck
CAS Number:
Specifications
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:96.11 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:75-75-2
Formula:CH₃SO₃H
MDL Number:MFCD00007518