Order Entry
Ireland
ContactUsLinkComponent
Rabbit Extendin-4 Acetate
Rabbit Extendin-4 Acetate
Supplier:  Cayman Chemical
Certificates
undefined
Rabbit Extendin-4 Acetate
Supplier:  Cayman Chemical

Specifications

  • CAS No.:
    914454-01-6
  • Species:
    Rabbit
  • Storage conditions:
    –20 °C
  • Protein synonyms:
    Exenatide acetate
  • Protein/peptide name:
    Extendin-4 Acetate
  • Purity:
    >95%
  • Molecular weight:
    4246,6
  • Formulation:
    Crystalline solid
  • Molecular formula:
    C₁₈₆H₂₈₆N₅₀O₆₂S
  • Shipping temperature:
    Ambient temperature

Specifications

Frequently Bought Together