Order Entry
Ireland
ContactUsLinkComponent
 
 
 
2-Propanol, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Reag. USP, Riedel-de Haën™
    
 
undefined
 
 
2-Propanol, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Reag. USP, Riedel-de Haën™
    

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion

 

 

  • Appearance:
    complying
  • Assay (GC):
    Min. 99.9 %
  • IR spectrum:
    complying
  • Boiling range:
    82.1 - 82.6 °C
  • Density (D 20/4):
    0.784 - 0.786
  • Refractive index (n 20/D):
    1.3770 - 1.3780
  • Non-volatile matter:
    Max. 10 ppm
  • Water (Karl Fischer):
    Max. 0.1 %
  • Free acid (as C₂H₅COOH):
    Max. 0.00074 %
  • Free alkali (as NH₃):
    Max. 0.00017 %
  • Free acid, free alkali:
    Max. 0.0001 meq/g
  • Peroxides (as H₂O₂):
    Max. 0.00034 %
  • Reaction against H₂SO₄:
    complying
  • KMnO₄ reducing matter (as O):
    Max. 5 ppm
  • Aluminium (Al):
    Max. 0.5 ppm
  • Boron (B):
    Max. 0.02 ppm
  • Barium (Ba):
    Max. 0.1 ppm
  • Bismuth (Bi):
    Max. 0.1 ppm
  • Calcium (Ca):
    Max. 0.5 ppm
  • Cadmium (Cd):
    Max. 0.01 ppm
  • Cobalt (Co):
    Max. 0.01 ppm
  • Chromium (Cr):
    Max. 0.01 ppm
  • Copper (Cu):
    Max. 0.01 ppm
  • Iron (Fe):
    Max. 0.1 ppm
  • Potassium (K):
    Max. 0.5 ppm
  • Lithium (Li):
    Max. 0.1 ppm
  • Magnesium (Mg):
    Max. 0.1 ppm
  • Manganese (Mn):
    Max. 0.01 ppm
  • Molybdenum (Mo):
    Max. 0.1 ppm
  • Sodium (Na):
    Max. 1 ppm
  • Nickel (Ni):
    Max. 0.01 ppm
  • Lead (Pb):
    Max. 0.01 ppm
  • Tin (Sn):
    Max. 0.1 ppm
  • Strontium (Sr):
    Max. 0.1 ppm
  • Zinc (Zn):
    Max. 0.01 ppm
  • Heavy metals (as Pb):
    Max. 1 ppm
  • Carbonyl compounds (as Acetone):
    Max. 20 ppm
  • Carbonyl compounds (as propionaldehyde):
    Max. 20 ppm
  • Ethanol (GC):
    Max. 0.01 %
  • Methanol (GC):
    Max. 0.1 %
  • APHA:
    Max. 10
  • Solubility in water:
    complying

 

 

Frequently Bought Together