


Nitric acid 65%, AR®, Macron Fine Chemicals™
Catalog # 3131-46
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Appearance:Clear, Colorless Or Yellowish Liquid
- Assay:≥65%
- Residue after Ignition:≤0.005%
- Chloride (Cl):≤0.0003%
- Phosphate (PO₄):≤0.001%
- Sulphate (SO₄):≤0.001%
- Heavy Metals (as Pb):≤0.0003%
- Arsenic (As):≤0.000003%
- Iron (Fe):≤0.0005%
- Lead (Pb):≤0.0001%
- Magnesium (Mg):≤0.0001%
- Manganese (Mn):≤0.00005%
Specification Test Results
About this item
:
:63.01 g/mol
UN:2031
ADR:8,II
:GHS03
:GHS05
:GHS06
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349