Sheepshead minnow (Cyprinodon variegatus), Proficiency Testing Material, Supelco®
Supplier: Merck
Certificates
About this item
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
pH Standard Solutions, IUPAC, Certified
Supplier: Hach