Sigma Pseudo™ Corpse Scent, Formulation II, Supelco®
Supplier: Merck
Certificates
About this item
Canine training aids for the detection of corpses. For post-putrification detection.
Sigma pseudo™ corpse scent may be used as an effective artificial scent in training cadaver dogs. The volatile compounds of its decomposition have been analyzed using gas chromatography coupled with time of flight mass spectrometry (GC–TOFMS).
Specifications
- Description:Sigma Pseudo(TM) Corpse Scent