Order Entry
Ireland
Orders LinkContactUsLinkComponent
 

Trichlorofluoromethane solution, NMR reference standard, Supelco®

Trichlorofluoromethane solution, 50% in chloroform-d (99.8 atom % D), NMR reference standard, NMR tube size 5 mm x 8 in., 1
Catalog # 612715-1EA
Supplier:   Merck
Trichlorofluoromethane solution, NMR reference standard, Supelco®
Catalog # 612715-1EA
Supplier:  Merck
CAS Number:  
Trichlorofluoromethane solution, NMR reference standard, Supelco®
Trichlorofluoromethane solution, NMR reference standard, Supelco®

Trichlorofluoromethane solution, 50% in chloroform-d (99.8 atom % D), NMR reference standard, NMR tube size 5 mm x 8 in., 1

CAS Number:  
Price
294,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications

Specifications

  • Concentration:
    50% in chloroform-d (99.8 atom % D)
  • Pk:
    1
  • Standards:
    NMR reference standard
  • Description:
    Trichlorofluoromethane solution
  • Pk Info:
    NMR tube size 5 mm x 8 in.
  • Cat. No.:
    612715-1EA
  • Description:
    Trichlorofluoromethane solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.