Order Entry
Ireland
ContactUsLinkComponent
Benzene solution, NMR reference standard, Supelco®
Benzene solution, NMR reference standard, Supelco®
Catalog # 611980-1EA
Supplier:  Merck
CAS Number:  
Benzene solution, NMR reference standard, Supelco®
Catalog # 611980-1EA
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    80% in acetone-d6 (99.9 atom % D)
  • Pk:
    1
  • Standards:
    NMR reference standard
  • Description:
    Benzene solution
  • Description:
    Benzene solution
  • Pk Info:
    NMR tube size 5 mm x 8 in.

Specifications

About this item