Order Entry
Ireland
ContactUsLinkComponent
Copper di-ammonium EDTA 0.1 mol/l, AVS TITRINORM
Copper di-ammonium EDTA 0.1 mol/l, AVS TITRINORM
Catalog # 86186.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
Copper di-ammonium EDTA 0.1 mol/l, AVS TITRINORM
Catalog # 86186.260
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • di-ammonium copper EDTA:
    0.1 mol/l
  • Appearance:
    clear
  • Colour:
    Blue
  • Density (20 °C):
    1,020 – 1,030 g/ml

Specification Test Results

About this item

 :
 :387.83 g/mol
CAS Number:67989-88-2
Formula:C₁₀H₂₀CuN₄O₈
MDL Number:MFCD01863036