Order Entry
Ireland
ContactUsLinkComponent
Mop heads, dry
Mop heads, dry
Catalog # MICRMZDM62-14
Supplier:  MICRONOVA
undefined
Mop heads, dry
Catalog # MICRMZDM62-14
Supplier:  MICRONOVA
Specifications
  • Description:
    Mop head, NovaPoly, Novalite
  • Length:
    356 mm
  • Seq:
    04
  • Pk:
    1
Specifications
About this item
Mops absorb detergent quickly and may be used on floors, walls, and ceilings without a bucket or wringer.
  • May be used with disinfectants
Material is designed for applications in which excess moisture is a concern. A special fabric strip allows for fast-drying action. Mop heads are sturdy enough to be laundered and reused. Latex/rayon blend mop head provides absorbency like that of PVA.