Order Entry
Ireland
ContactUsLinkComponent
 
Organic reference standard, Undecanal, analytical standard, Supelco®
  80139-1ML
 :  Merck
 :  
undefined
Organic reference standard, Undecanal, analytical standard, Supelco®
  80139-1ML
 :  Merck
 :  

 

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    112-44-7
  • Description:
    Undecanal, ≥98,0% (GC)
  • Melting point:
    −2 °C (lit.)
  • Formula:
    C₁₁H₂₂O
  • Description:
    Undecanal
  • Boiling point:
    109...115 °C/5 mm Hg (lit.)
  • Form:
    Neat