Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Trimethylamine hydrochloride, analytical standard, Supelco®
Organic reference standard, Trimethylamine hydrochloride, analytical standard, Supelco®
Catalog # 41284-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Trimethylamine hydrochloride, analytical standard, Supelco®
Catalog # 41284-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    593-81-7
  • Description:
    Trimethylamine hydrochloride, ≥98,5% (GC)
  • Melting point:
    283...284 °C (dec.) (lit.)
  • Formula:
    C₃H₁₀ClN
  • Description:
    Trimethylamine hydrochloride

Specifications

About this item