Organic reference standard, trans-3-Octen-2-one, analytical standard, Supelco®
Catalog # 41339-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:18402-82-9
- Description:trans-3-Octen-2-one, ≥98,0% (GC)
- Formula:C₈H₁₄O
- Description:trans-3-Octen-2-one
- Boiling point:100 °C/18 mm Hg (lit.)