Organic reference standard, trans-2,cis-6-Nonadienal, analytical standard, Supelco®
Catalog # 05549-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:557-48-2
- Description:trans-2,cis-6-Nonadienal, ≥94,5% (GC)
- Formula:C₉H₁₄O
- Description:trans-2,cis-6-Nonadienal
- Boiling point:94...95 °C/18 mm Hg (lit.)