Order Entry
Ireland
ContactUsLinkComponent
 
Organic reference standard, Sucralose, analytical standard, Supelco®
Catalog # 90984-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Sucralose, analytical standard, Supelco®
Catalog # 90984-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    56038-13-2
  • Description:
    Sucralose, ≥98,0% (HPLC)
  • Formula:
    C₁₂H₁₉Cl₃O₈
  • Description:
    Sucralose
  • Form:
    Neat

Specifications

About this item

Sucralose is a derivative of sucrose, which is generally used as a non-nutritive sweetener in foods and beverages.

A synthetic sweet tastant detectable by humans. Activates T1R2/T1R3 sweet taste receptors on enteroendocrine cells and elicits increased hormonal secretion of glucagon-like peptide-1 and glucose-dependent insulinotrophic peptide.

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.