Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Stevioside, analytical standard, Supelco®
Organic reference standard, Stevioside, analytical standard, Supelco®
Catalog # 50956-10MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Stevioside, analytical standard, Supelco®
Catalog # 50956-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    57817-89-7
  • Description:
    Stevioside, ≥95,0% (HPLC)
  • Formula:
    C₃₈H₆₀O₁₈
  • Description:
    Stevioside
  • Form:
    Neat

Specifications

About this item

Stevioside is a diterpene glycoside, naturally extracted from the leaves of Stevia rebaudiana Bertoni plant, which can find wide applications as a commercial non-caloric sweetener and can also be used in the treatment of diabetes.