Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Sterigmatocystin, Supelco®
Organic reference standard, Sterigmatocystin, Supelco®
Catalog # 32609-5MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Sterigmatocystin, Supelco®
Catalog # 32609-5MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mg
  • CAS No.:
    10048-13-2
  • Description:
    Sterigmatocystin
  • Formula:
    C₁₈H₁₂O₆
  • Storage temperature:
    −20 °C
  • Description:
    Sterigmatocystin
  • Form:
    Neat

Specifications

About this item

Sterigmatocystin is classified under the mycotoxin family of compounds that is usually produced by fungi of several species of Aspergillus.

Initiates lung and liver tumors under experimental conditions.

Sterigmatocystin may be used as an analytical reference standard for the determination of the analyte in, Grain samples by high-performance liquid chromatography-electrospray positive ionization-tandem mass spectrometry (HPLC-ES⁺ MS/MS). Sweet pepper samples by LC-ESI⁺ MS/MS.