Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Spermidine, analytical standard, Supelco®
Organic reference standard, Spermidine, analytical standard, Supelco®
Catalog # 49761-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Spermidine, analytical standard, Supelco®
Catalog # 49761-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    124-20-9
  • Description:
    Spermidine, ≥97.5% (NT), ≥99,0% (GC)
  • Formula:
    C₅H₁₀O₂
  • Storage temperature:
    2...8°C
  • Description:
    Spermidine
  • Form:
    Neat

Specifications

About this item

Spermidine is a naturally occurring polyamine compound commonly found in food. It is essential for cell growth and survival, technique. Contact Technical Service for further support.

Inhibits neuronal nitric oxide synthase (nNOS). Binds and precipitates DNA, may be used for purification of DNA binding proteins. Stimulates T4 polynucleotide kinase activity.

Solutions should be sterile-filtered, not autoclaved, if sterile solution is necessary. Spermidine deaminates with time, solutions should be stored frozen. Prepare new solutions frequently.

Spermidine may be used as one of the reference standard for the determination of spermidine in biological samples by high performance liquid chromatography-electrospray ionisation–ion trap tandem mass spectrometry (HPLC–ESI–ITMS/MS).

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.