Organic reference standard, Phthalic acid, analytical standard, Supelco®
79255-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:88-99-3
- Description:Phthalic acid, ≥99,5% (GC) and ≥99,5% (T)
- Melting point:210...211 °C (lit.)
- Formula:C₈H₆O₂
- Description:Phthalic acid
- Form:Neat