Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Patulin solution, analytical standard, Supelco®
Organic reference standard, Patulin solution, analytical standard, Supelco®
Catalog # 34127-2ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Patulin solution, analytical standard, Supelco®
Catalog # 34127-2ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml in acetonitrile
  • Pk:
    2 mL
  • Standards:
    Analytical standard
  • CAS No.:
    149-29-1
  • Description:
    Patulin solution
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₇H₆O₄
  • Storage temperature:
    –20 °C
  • Description:
    Patulin solution

Specifications

About this item

Patulin is an important mycotoxin produced by several species of mold, especially within Penicillium, Aspergillus and Byssochlamys. It is commonly found in fruits especially apples, pears and grapes.

Patulin is a mycotoxin produced by several mold species. It has been found in fruit, especially apples, fruit juice, cider and stewed fruits. Because of its highly toxic properties, this compound has been the focus of many studies.

Patulin solution may be used as an analytical reference standard for the determination of the analyte in dried apple rings using high-performance liquid chromatography with diode array detection (HPLC-DAD) and fruit juices using high-performance liquid chromatography with diode array detection (HPLC-DAD) and gas chromatography coupled to mass spectrometry (GC-MS).