Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Nodularin solution, analytical standard, Supelco®
Organic reference standard, Nodularin solution, analytical standard, Supelco®
Catalog # 32539-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Nodularin solution, analytical standard, Supelco®
Catalog # 32539-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10 µg/ml in methanol
  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    118399-22-7
  • Description:
    Nodularin solution
  • Formula:
    C₄₁H₆₀N₈O₁₀
  • Storage temperature:
    –20 °C
  • Description:
    Nodularin solution
  • Flash point:
    11 °C

Specifications

About this item

Nodularins belong to the class of hepatotoxic cyclic pentapeptides and have their structures resembling to that of microcystins. They are produced by Nodularia spumigena cyanobacterium.

Nodularin may be used as an analytical reference standard for the determination of the analyte in: surface and drinking waters by on-line solid phase extraction (SPE) and ultra high pressure liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) and Sediments and pore waters by accelerated solvent extraction (ASE) and HPLC-MS/MS.