Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Nerol, analytical standard, Supelco®
Organic reference standard, Nerol, analytical standard, Supelco®
  50949-1ML
 :  Merck
 :  
undefined
Organic reference standard, Nerol, analytical standard, Supelco®
  50949-1ML
 :  Merck
 :  

 

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    106-25-2
  • Description:
    Nerol, ≥97,0% (GC)
  • Formula:
    C₁₀H₁₈O
  • Description:
    Nerol
  • Flash point:
    107 °C
  • Boiling point:
    103…105 °C/9 mmHg
  • Form:
    Neat

 

 

Nerol belongs to the class of monoterpene glycosides.

Nerol may be used as an analytical reference standard for the determination of nerol in: e-cigarette refill solutions by high performance liquid chromatography-tandem mass spectrometry with electrospray ionization (HPLC-ESI-MS/MS) as well as gas chromatography (GC) coupled to MS/MS operating under the electron impact (EI) mode and multiple reaction monitoring (MRM) mode of detection.

Honeybush tea (Cyclopia spp.) by headspace solid phase microextraction (HS-SPME) and comprehensive two-dimensional gas chromatography (GC x GC) with flame ionization detection (FID) and a single stage modulator. Hanzhong black tea (Camellia sinesis) by solid phase extraction (SPE) and GC coupled to MS as well as olfactometry.

Wine by liquid-liquid extraction (LLE) and EI-GC-MS with selective ion monitoring (SIM) detection.