Organic reference standard, Neosolaniol, Supelco®
32932-5MG
: Merck
:
- Pk:5 mg
- CAS No.:36519-25-2
- Description:Neosolaniol
- M-Clarity quality level:MQ200
- Formula:C₁₉H₂₆O₈
- Storage temperature:−20 °C
- Description:Neosolaniol
- Form:Neat
Neosolaniol belongs to the class of type A trichothecene mycotoxins produced by various Fusarium molds.
Neosolaniol may be used as an analytical reference standard for the determination of the analyte in: Wheat samples by acetonitrile-based extraction and gas chromatography-tandem mass spectrometry (GC-MS/MS) as well as capillary GC with flame ionization detection and Cereals by acetonitrile-based extraction and GC coupled to mass spectrometry (MS).