Order Entry
Ireland
ContactUsLinkComponent
 
Organic reference standard, Naphthol Yellow S, analytical standard, Supelco®
Catalog # 49547-25MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Naphthol Yellow S, analytical standard, Supelco®
Catalog # 49547-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    846-70-8
  • Description:
    Naphthol Yellow S, ≥99,0% (HPLC)
  • Formula:
    C₁₀H₄N₂Na₂O₈S
  • Description:
    Naphthol Yellow S
  • Form:
    Neat

Specifications

About this item

Naphthol Yellow S is a dye that is known for its hazardous nature.

Naphthol Yellow S may be used as an analytical reference standard for the determination of Naphthol Yellow S in: Foodstuffs by solid phase extraction (SPE) and high performance liquid chromatography (HPLC)-diode array detection (DAD)-ion trap time-of-flight tandem mass spectrometry (IT-TOF-MS3) equipped with multiple reaction monitoring (MRM) detection.

Drinks and candies by SPE followed by analysis using HPLC coupled to photodiode array (PDA) detector. Wastewater samples by adsorption column chromatography and embroideries designed by the famous 19th century French painter Emile Bernard.

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.