Order Entry
Ireland
ContactUsLinkComponent
 
Organic reference standards, Monosaccharides Kit, (individually packaged in quantities indicated), analytical standard, Supelco®
Catalog # 47267.
Supplier:  Merck
CAS Number:  
Organic reference standards, Monosaccharides Kit, (individually packaged in quantities indicated), analytical standard, Supelco®
Catalog # 47267.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    Monosaccharides kit
  • Storage temperature:
    2…8 °C
  • Pk Info:
    1 kit
  • Form:
    Neat

Specifications

About this item

Kit components:

D-(+)Glucose, mixed anomers (47249) 500 mg; D-(-)Arabinose 500 mg; Fructose 500 mg; D-(+)Galactose 500 mg; D-(+)Mannose (mixed anomers) 500 mg; D-(-)Ribose 500 mg; D-(+)Xylose 500 mg