Organic reference standard, Melamine-13C3, analytical standard, VETRANAL™, Supelco®
32666-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:1173022-88-2
- Description:Melamine-13C3
- Melting point:>300 °C
- Formula:13C₃H₆N₆
- Description:Melamine-13C3
- Flash point:300 °C
- Form:Neat
Melamine-¹³C₃ may be used as an analytical standard for the determination of melamine in milk powder samples by liquid chromatography-electrospray ionisation-isotope dilution mass spectrometry (LC-ESI-IDMS).