Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Lauric aldehyde, analytical standard, Supelco®
Organic reference standard, Lauric aldehyde, analytical standard, Supelco®
Catalog # 55977-5ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Lauric aldehyde, analytical standard, Supelco®
Catalog # 55977-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    112-54-9
  • Description:
    Lauric aldehyde, ≥98,0% (GC)
  • Seq:
    02
  • Formula:
    C₁₂H₂₄O
  • Description:
    Lauric aldehyde
  • Boiling point:
    185 °C/100 mm Hg (lit.)
  • Form:
    Neat

Specifications

About this item