Organic reference standard, Hexyl salicylate, analytical standard, Supelco®
Catalog # 61690-5ML
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mL
- Standards:Analytical standard
- CAS No.:6259-76-3
- Description:Hexyl salicylate, ≥99,0% (GC)
- Formula:C₁₃H₁₈O₃
- Description:Hexyl salicylate