Order Entry
Ireland
ContactUsLinkComponent
 
Organic reference standard, Heptanoic acid, analytical standard, Supelco®
 :  Merck
 
undefined
Organic reference standard, Heptanoic acid, analytical standard, Supelco®
 :  Merck
 

 

  • CAS No.:
    111-14-8
  • Melting point:
    −10,5 °C (lit.)
  • Formula:
    C₇H₁₄O₂
  • Description:
    Heptanoic acid
  • Boiling point:
    223 °C (lit.)

 

 

Frequently Bought Together