Order Entry
Ireland
ContactUsLinkComponent
 
Organic reference standard, Furfuryl alcohol, analytical standard, Supelco®
 :  Merck
 
undefined
Organic reference standard, Furfuryl alcohol, analytical standard, Supelco®
 :  Merck

 

  • CAS No.:
    98-00-0
  • Melting point:
    −29 °C
  • Formula:
    C₅H₆O₂
  • Description:
    Furfuryl alcohol, ≥97,5% (GC)
  • Boiling point:
    170 °C

 

Frequently Bought Together