Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, ethyl palmitate, analytical standard, Supelco®
Organic reference standard, ethyl palmitate, analytical standard, Supelco®
  67107-250MG
 :  Merck
 :  
undefined
Organic reference standard, ethyl palmitate, analytical standard, Supelco®
  67107-250MG
 :  Merck
 :  

 

  • Pk:
    250 mg
  • Standards:
    Analytical standard
  • CAS No.:
    628-97-7
  • Description:
    Ethyl palmitate, ≥98,5% (GC)
  • Melting point:
    24...26 °C (lit.)
  • Formula:
    C₁₈H₃₆O₂
  • Description:
    Ethyl palmitate
  • Boiling point:
    192-193 °C/10 mmHg (lit.)
  • Form:
    Neat