Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, ethyl maltol, analytical standard, Supelco®
Organic reference standard, ethyl maltol, analytical standard, Supelco®
Catalog # 94185-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, ethyl maltol, analytical standard, Supelco®
Catalog # 94185-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    4940-11-8
  • Description:
    Ethyl maltol, ≥98,0% (GC)
  • Melting point:
    85...95 °C (lit.)
  • Formula:
    C₇H₈O₃
  • Description:
    Ethyl maltol
  • Form:
    Neat

Specifications

About this item

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.