Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Ethyl benzoate, analytical standard, Supelco®
Organic reference standard, Ethyl benzoate, analytical standard, Supelco®
Supplier:  Merck
Certificates
undefined
Organic reference standard, Ethyl benzoate, analytical standard, Supelco®
Supplier:  Merck

Specifications

  • CAS No.:
    93-89-0
  • Melting point:
    −34 °C (lit.)
  • Formula:
    C₉H₁₀O₂
  • Description:
    Ethyl benzoate
  • Boiling point:
    212 °C (lit.)

Specifications

Frequently Bought Together