Order Entry
Ireland
ContactUsLinkComponent
Supelco® Organic Reference Standard, ᴅ-Pinitol
Supelco® Organic Reference Standard, ᴅ-Pinitol
Catalog # 74948-25MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, ᴅ-Pinitol
Catalog # 74948-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    10284-63-6
  • Description:
    ᴅ-Pinitol
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00216659
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3-O-Methyl-ᴅ-chiro-inositol
  • Melting point:
    179 - 185 °C (lit.)
  • Molecular weight:
    194,18

Specifications

About this item