Organic reference standard, delta-Octalactone, mixture of isomers, analytical standard, Supelco®
Catalog # 94490-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:698-76-0
- Description:delta-Octalactone, mixture of isomers, ≥98,0% (GC)
- Seq:1
- Formula:C₈H₁₄O₂
- Description:delta-Octalactone
- Flash point:110 °C
- Form:Neat