Organic reference standard, delta-Dodecalactone, analytical standard, Supelco®
92255-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:713-95-1
- Description:delta-Dodecalactone, ≥98,0% (GC)
- Melting point:–12 °C (lit.)
- Formula:C₁₂H₂₂O₂
- Description:delta-Dodecalactone
- Boiling point:140...141 °C/1 mmHg (lit.)
- Form:Neat
: Bottomless glass bottle. Contents are inside inserted fused cone.