Organic reference standard, delta-Decalactone, analytical standard, Supelco®
Catalog # 74026-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:705-86-2
- Description:delta-Decalactone, ≥98,0% (GC)
- Melting point:–27 °C (lit.)
- Formula:C₁₀H₁₈O₂
- Description:delta-Decalactone
- Boiling point:117...120 °C/0,02 mmHg (lit.)