Organic reference standard, Bisphenol BP, analytical standard, Supelco®
08138-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:1844-01-5
- Description:Bisphenol BP, ≥98,0% (HPLC)
- Formula:C₂₅H₂₀O₂
- Description:Bisphenol BP
Bisphenol BP belongs to the class of bisphenols.
Bisphenol BP may be used as a reference standard in the determination of bisphenol BP in serum samples using high performance liquid chromatography coupled with tandem mass spectrometry (HPLC-MS/MS).