Order Entry
Ireland
ContactUsLinkComponent
 
Organic reference standard, Bisphenol B, analytical standard, Supelco®
Catalog # 50877-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Bisphenol B, analytical standard, Supelco®
Catalog # 50877-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    77-40-7
  • Description:
    Bisphenol B, ≥98,0% (HPLC)
  • Formula:
    C₁₆H₁₈O₂
  • Description:
    Bisphenol B

Specifications

About this item

Bisphenol B is a congener of bisphenol A, used primarily for the manufacturing of phenolic resins and is also a potent food contaminant.

Bisphenol B may be used as a reference standard for the determination of bisphenol B in liquid food matrices and powdered infant formulae and human urine samples by using dispersive liquid-liquid microextraction (DLLME) with in situ derivatisation coupled with heart-cutting gas chromatography-mass spectrometry (GC-MS).