Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Bisphenol A bis(2,3-dihydroxypropyl) ether, analytical standard, Supelco®
Organic reference standard, Bisphenol A bis(2,3-dihydroxypropyl) ether, analytical standard, Supelco®
Catalog # 15137-250MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Bisphenol A bis(2,3-dihydroxypropyl) ether, analytical standard, Supelco®
Catalog # 15137-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    Analytical standard
  • CAS No.:
    5581-32-8
  • Description:
    Bisphenol A bis(2,3-dihydroxypropyl) ether, ≥97,0% (HPLC)
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₂₁H₂₈O₆
  • Description:
    Bisphenol A bis(2,3-dihydroxypropyl) ether
  • Form:
    Neat

Specifications

About this item

Standard for determining toxic monomers released from polymers of the inner coating of cans.

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.