Order Entry
Ireland
ContactUsLinkComponent
 
Organic reference standard, Bisphenol A (3-chloro-2-hydroxypropyl) glycidyl ether, analytical standard, Supelco®
  73124-100MG
 :  Merck
 :  
undefined
Organic reference standard, Bisphenol A (3-chloro-2-hydroxypropyl) glycidyl ether, analytical standard, Supelco®
  73124-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    13836-48-1
  • Description:
    Bisphenol A (3-chloro-2-hydroxypropyl) glycidyl ether, ≥90% (HPLC)
  • Seq:
    02
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₂₁H₂₅ClO₄
  • Storage temperature:
    –20 °C
  • Description:
    Bisphenol A (3-chloro-2-hydroxypropyl) glycidyl ether
  • Form:
    Neat

 

 

Bisphenol A (3-chloro-2-hydroxypropyl) glycidyl ether is a chlorinated derivative, which is released during the thermal coating treatment of food cans, big storage vessels and food containers.

Bisphenol A (3-chloro-2-hydroxypropyl) glycidyl ether may be used as a reference standard for the determination of bisphenol A (3-chloro-2-hydroxypropyl) glycidyl ether in canned food and beverages using fast liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS).

 : Bottomless glass bottle. Contents are inside inserted fused cone.