Order Entry
Ireland
ContactUsLinkComponent
 
Organic reference standard, Bis(2-ethylhexyl) terephthalate, analytical standard, Supelco®
Catalog # 49234-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Bis(2-ethylhexyl) terephthalate, analytical standard, Supelco®
Catalog # 49234-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    6422-86-2
  • Description:
    Bis(2-ethylhexyl) terephthalate, ≥97,0% (GC)
  • Melting point:
    –48 °C
  • Formula:
    C₂₄H₃₈O₄
  • Description:
    Bis(2-ethylhexyl) terephthalate
  • Boiling point:
    400 °C

Specifications

About this item