Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, beta-Cyclocitral, analytical standard, Supelco®
Organic reference standard, beta-Cyclocitral, analytical standard, Supelco®
Catalog # 16976-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, beta-Cyclocitral, analytical standard, Supelco®
Catalog # 16976-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    432-25-7
  • Description:
    beta-Cyclocitral, ≥97,0% (GC)
  • Formula:
    C₁₀H₁₆O
  • Storage temperature:
    2...8 °C
  • Description:
    beta-Cyclocitral
  • Boiling point:
    62...63 °C/3 mmHg (lit.)

Specifications

About this item