Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Benzyl salicylate, analytical standard, Supelco®
Organic reference standard, Benzyl salicylate, analytical standard, Supelco®
Catalog # 51031-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Benzyl salicylate, analytical standard, Supelco®
Catalog # 51031-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    118-58-1
  • Description:
    Benzyl salicylate, ≥98,5% (GC)
  • Formula:
    C₁₄H₁₂O₃
  • Description:
    Benzyl salicylate
  • Boiling point:
    168...170 °C/5 mmHg (lit.)
  • Form:
    Neat

Specifications

About this item