Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Benzyl cyanide, analytical standard, Supelco®
Organic reference standard, Benzyl cyanide, analytical standard, Supelco®
Catalog # 12608-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Benzyl cyanide, analytical standard, Supelco®
Catalog # 12608-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    140-29-4
  • Description:
    Benzyl cyanide, ≥98,0% (GC)
  • Melting point:
    –24 °C
  • Formula:
    C₈H₇N
  • Description:
    Benzyl cyanide
  • Boiling point:
    233...234 °C (lit.)

Specifications

About this item