Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, alpha-Damascone, analytical standard, Supelco®
Organic reference standard, alpha-Damascone, analytical standard, Supelco®
Catalog # 59574-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, alpha-Damascone, analytical standard, Supelco®
Catalog # 59574-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    43052-87-5
  • Description:
    alpha-Damascone, ≥95,0% (GC)
  • Formula:
    C₁₃H₂₀O
  • Description:
    alpha-Damascone

Specifications

About this item