Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Aflatoxin G₂-¹³C₁₇ solution, analytical standard, Supelco®
Organic reference standard, Aflatoxin G₂-¹³C₁₇ solution, analytical standard, Supelco®
Catalog # 32777-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Aflatoxin G₂-¹³C₁₇ solution, analytical standard, Supelco®
Catalog # 32777-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    0,5 µg/ml in acetonitrile
  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    1217462-49-1
  • Description:
    Aflatoxin G2-13C17 solution
  • Formula:
    13C₁₇H₁₄O₇
  • Storage temperature:
    –20 °C
  • Description:
    Aflatoxin G2-13C17 solution
  • Form:
    Neat

Specifications

About this item

Aflatoxin G₂-¹³C₁₇ is the ¹³C isotope-labeled analog of the mycotoxin, aflatoxin.

  • Analytical standard for the determination of aflatoxin G₂ in food grade gums as well as milk, milk-based infant formula

Feed samples are stable isotope dilution and liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).