Specifications
- Concentration:0.5 µg/ml in acetonitrile
- Pk:2 mL
- Standards:Analytical standard
- CAS No.:7220-81-7
- Description:Aflatoxin B₂ solution
- M-Clarity quality level:MQ100
- Formula:C₁₇H₁₄O₆
- Storage temperature:−20 °C
- Description:Aflatoxin B₂ solution
- Flash point:2 °C
Specifications
About this item
Aflatoxins are mycotoxins that are generated as a result of secondary metabolism of various fungal species such as Aspergillus, Penicillium and Fusarium.
Aflatoxin B₂ may be used for the determination of aflatoxin B₂ in: Animal feed by high performance liquid chromatography with fluorescence detection (HPLC-FLD) as well as quick, easy, cheap, effective, rugged and safe (QuEchERS) extraction method combined with ultra-high performance liquid chromatography-high resolution mass spectrometry (UHPLC-HRMS). Eggs by QuEchERS extraction method followed by analysis using UHPLC with tandem mass spectrometry (MS/MS) operating on electrospray-ionization interface (ESI) mode. Brazil nuts by immunoaffinity column (IAC) and clean-up HPLC-FLD. Plant-based milk and dairy products by dispersive liquid-liquid microextraction (DLLME), QuEchERS extraction method and HPLC-FLD.